Learn More
Medchemexpress LLC L-Ascorbic acid | 50-81-7 | MFCD00064328 | 99.97% | 50 G
SDP
Catalog No. 5000406966
Encompass_Preferred
L-Ascorbic acid is an electron donor and an endogenous antioxidant agent. It selectively inhibits Cav3.2 channels with an IC50 of 6.5 μM. Additionally, it acts as a collagen deposition enhancer, an elastogenesis inhibitor, and demonstrates anti-cancer effects by generating reactive oxygen species (ROS) to selectively damage cancer cells.
  • Electron donor and endogenous antioxidant agent
  • Selectively inhibits Cav3.2 channels
  • Enhances collagen deposition
  • Inhibits elastogenesis
  • Demonstrates anti-cancer effects
  • Ships at room temperature in continental US
  • Solid form stored at room temperature, protected from light
  • Solution form stored at -80°C for 1 year, or -20°C for 6 months
Catalog No. 50-004-06966 Supplier Medchemexpress LLC Supplier No. HYB016650G
L-Ascorbic acid is an electron donor and an endogenous antioxidant agent. It selectively inhibits Cav3.2 channels with an IC50 of 6.5 μM. Additionally, it acts as a collagen deposition enhancer, an elastogenesis inhibitor, and demonstrates anti-cancer effects by generating reactive oxygen species (ROS) to selectively damage cancer cells.
  • Electron donor and endogenous antioxidant agent
  • Selectively inhibits Cav3.2 channels
  • Enhances collagen deposition
  • Inhibits elastogenesis
  • Demonstrates anti-cancer effects
  • Ships at room temperature in continental US
  • Solid form stored at room temperature, protected from light
  • Solution form stored at -80°C for 1 year, or -20°C for 6 months