Learn More
Cayman Chemical L-Ascorbic Acid, Purity: ≥95%, CAS: 50-81-7, 50 g
SDP
Catalog No. 502968728
Encompass_Preferred
A naturally occurring electron donor for eight different enzymes, including those related to collagen hydroxylation, carnitine synthesis (which aids in the generation of ATP), norepinephrine synthesis, tyrosine metabolism, and amidating peptides
Catalog No. 50-296-8728 Supplier Cayman Chemical Supplier No. 1465650 G
A naturally occurring electron donor for eight different enzymes, including those related to collagen hydroxylation, carnitine synthesis (which aids in the generation of ATP), norepinephrine synthesis, tyrosine metabolism, and amidating peptides