Learn More
Cayman Chemical L-Ascorbic Acid, 50-81-7, 50 g
SDP
Supplier:  Cayman Chemical 1465650
Encompass_Preferred
A naturally occurring electron donor for eight different enzymes, including those related to collagen hydroxylation, carnitine synthesis (which aids in the generation of ATP), norepinephrine synthesis, tyrosine metabolism, and amidating peptides
Catalog No. NC1343993