Learn More
Sigma Aldrich Fine Chemicals Biosciences L Ascorbic acid 99
Catalog No. 502903783
Encompass
L-ascorbic acid is used as a food additive browning inhibitor flavor stabilizer dough modifier and color stabilizer. Deficiency of ascorbic acid is associated with scurvy. L-ascorbic acid stimulates immune response and is implicated in the synthesis of collagen carnitine and neurotransmitters.
Catalog No. 50-290-3783 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A929022KG
L-ascorbic acid is used as a food additive browning inhibitor flavor stabilizer dough modifier and color stabilizer. Deficiency of ascorbic acid is associated with scurvy. L-ascorbic acid stimulates immune response and is implicated in the synthesis of collagen carnitine and neurotransmitters.