Learn More
Sigma Aldrich Fine Chemicals Biosciences L Ascorbic acid BioXtra 99500G
Catalog No. 502903834
Encompass
Ascorbic acid is also referred to as Vitamin C. It contains a six-carbon lactone produced by plants and some animal species but not by humans and other primates. Vitamin C is a water-soluble vitamin. Vitamin C is a part of citrus fruits such as Indian gooseberry broccoli strawberries brussels sprouts. L-ascorbic acid is the biologically active form of ascorbic acid. It is unstable and hydrophilic. Ascorbic acid carries a neutral charge which gets converted to ascorbate by protonation.
Catalog No. 50-290-3834 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A5960500G
Ascorbic acid is also referred to as Vitamin C. It contains a six-carbon lactone produced by plants and some animal species but not by humans and other primates. Vitamin C is a water-soluble vitamin. Vitamin C is a part of citrus fruits such as Indian gooseberry broccoli strawberries brussels sprouts. L-ascorbic acid is the biologically active form of ascorbic acid. It is unstable and hydrophilic. Ascorbic acid carries a neutral charge which gets converted to ascorbate by protonation.