Learn More
Medchemexpress LLC L-ascorbic acid-d2 | 82977-10-4 | 99.9% | 178.14 | C6H6D2O6 | 10 MG
SDP
Catalog No. 5000296825
Encompass_Preferred
L-ascorbic acid-d2 is the deuterium-labeled form of L-ascorbic acid used as an analytical standard and tracer in biochemical and mass spectrometry applications. It is a white to off-white solid with molecular formula C6H6D2O6 and molecular weight 178.14 g/mol. Supplied in small mass pack sizes for analytical workflows.
  • Deuterium-labeled compound for tracer and quantitative analyses.
  • High chemical purity (99.9%) suitable for analytical use.
  • White to off-white solid, easy to handle and weigh.
  • Storable as powder at -20°C for long-term stability.
  • Available in small pack sizes for calibration and method development.
Catalog No. 50-002-96825 Supplier Medchemexpress LLC Supplier No. HYB0166S610MG
L-ascorbic acid-d2 is the deuterium-labeled form of L-ascorbic acid used as an analytical standard and tracer in biochemical and mass spectrometry applications. It is a white to off-white solid with molecular formula C6H6D2O6 and molecular weight 178.14 g/mol. Supplied in small mass pack sizes for analytical workflows.
  • Deuterium-labeled compound for tracer and quantitative analyses.
  • High chemical purity (99.9%) suitable for analytical use.
  • White to off-white solid, easy to handle and weigh.
  • Storable as powder at -20°C for long-term stability.
  • Available in small pack sizes for calibration and method development.