Learn More
Medchemexpress LLC L-Ascorbic acid-d2 | 82977-10-4 | 99.9% | 25 MG
SDP
Catalog No. 5000450978
Encompass_Preferred
L-Ascorbic acid-d2 is the deuterium labeled form of L-Ascorbic acid, an electron donor and endogenous antioxidant agent. It inhibits selectively Cav3.2 channels with an IC50 of 6.5 μM. This product is suitable for research use only and not intended for human use.
  • Deuterium labeled L-Ascorbic acid
  • Functions as an electron donor
  • Acts as an endogenous antioxidant agent
  • Inhibits selectively Cav3.2 channels
  • Can be used as a tracer
  • Useful as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-50978 Supplier Medchemexpress LLC Supplier No. 25MGHYB0166S6
L-Ascorbic acid-d2 is the deuterium labeled form of L-Ascorbic acid, an electron donor and endogenous antioxidant agent. It inhibits selectively Cav3.2 channels with an IC50 of 6.5 μM. This product is suitable for research use only and not intended for human use.
  • Deuterium labeled L-Ascorbic acid
  • Functions as an electron donor
  • Acts as an endogenous antioxidant agent
  • Inhibits selectively Cav3.2 channels
  • Can be used as a tracer
  • Useful as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS