Learn More
Sigma Aldrich Fine Chemicals Biosciences L Ascorbic acid FCC FG25KG
Catalog No. 502906943
Encompass
L-Ascorbic acid an organic acid is found abundantly in citrus fruits. It is generally used as an antioxidant and pH adjuster in the cosmetic industry. L-AA is also used as a preservative and nutrient supplement in foods.
Catalog No. 50-290-6943 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W21090125KG
L-Ascorbic acid an organic acid is found abundantly in citrus fruits. It is generally used as an antioxidant and pH adjuster in the cosmetic industry. L-AA is also used as a preservative and nutrient supplement in foods.