Learn More
Medchemexpress LLC L-Ascorbic acid (GMP Like) | 50-81-7 | MFCD00064328 | 99.9% | 50 MG
SDP
Catalog No. 5000406967
Encompass_Preferred
L-Ascorbic acid (GMP Like) is a GMP Like class of L-Ascorbic acid. Also known as L-Ascorbate or Vitamin C, it functions as an electron donor and an endogenous antioxidant agent. It selectively inhibits Cav3.2 channels with an IC50 of 6.5 μM and acts as a collagen deposition enhancer and an elastogenesis inhibitor. It also demonstrates anti-cancer effects by generating reactive oxygen species (ROS) and selectively damaging cancer cells.
  • Gmp like class L-Ascorbic acid.
  • Endogenous antioxidant agent.
  • Selectively inhibits Cav3.2 channels.
  • Enhances collagen deposition.
  • Inhibits elastogenesis.
  • Exhibits anti-cancer effects through ROS generation and selective cancer cell damage.
Catalog No. 50-004-06967 Supplier Medchemexpress LLC Supplier No. HYB0166GL50MG
L-Ascorbic acid (GMP Like) is a GMP Like class of L-Ascorbic acid. Also known as L-Ascorbate or Vitamin C, it functions as an electron donor and an endogenous antioxidant agent. It selectively inhibits Cav3.2 channels with an IC50 of 6.5 μM and acts as a collagen deposition enhancer and an elastogenesis inhibitor. It also demonstrates anti-cancer effects by generating reactive oxygen species (ROS) and selectively damaging cancer cells.
  • Gmp like class L-Ascorbic acid.
  • Endogenous antioxidant agent.
  • Selectively inhibits Cav3.2 channels.
  • Enhances collagen deposition.
  • Inhibits elastogenesis.
  • Exhibits anti-cancer effects through ROS generation and selective cancer cell damage.