Learn More
Medchemexpress LLC L-Ascorbic acid (L-Ascorbate) (GMP) | 50-81-7 | MFCD00064328 | 100 MG
SDP
Catalog No. 5000406968
Encompass_Preferred
L-Ascorbic acid (L-Ascorbate) (GMP) is Ascorbic acid produced using GMP guidelines. This GMP small molecule functions as an auxiliary reagent for cell therapy manufacture and acts as an inhibitor of Cav 3.2 channels.
  • Produced using GMP guidelines.
  • Functions as an auxiliary reagent for cell therapy manufacture.
  • Acts as an inhibitor of Cav 3.2 channels.
  • Induces chondrogenic differentiation of adipose-derived mesenchymal stem cells.
  • Induces osteogenic differentiation of mesenchymal stem cells (MSCs).
Catalog No. 50-004-06968 Supplier Medchemexpress LLC Supplier No. HYB0166G100MG
L-Ascorbic acid (L-Ascorbate) (GMP) is Ascorbic acid produced using GMP guidelines. This GMP small molecule functions as an auxiliary reagent for cell therapy manufacture and acts as an inhibitor of Cav 3.2 channels.
  • Produced using GMP guidelines.
  • Functions as an auxiliary reagent for cell therapy manufacture.
  • Acts as an inhibitor of Cav 3.2 channels.
  • Induces chondrogenic differentiation of adipose-derived mesenchymal stem cells.
  • Induces osteogenic differentiation of mesenchymal stem cells (MSCs).