Learn More
FUJIFILM Biosciences L-Ascorbic Acid Phosphate Mag
Catalog No. 5000450238
Encompass_Preferred
L-Ascorbic Acid Phosphate has been shown to promote cell growth and collagen synthesis Specifically Asc 2-P increases the steady state levels of mRNA for type I collagen chains and elevates the transcriptional rates of type I collagen genes Problems exist however with typical ascorbic acid the greatest of which is its short biological half-life Free ascorbic acid is very unstable in solution especially under culture conditions of neutral pH and 37 degrees C L-Ascorbic acid phosphate is much more stable product and can thus be used more easily and effectively Water-soluble high-purity form of Vitamin C phosphate For research use only RUO
Catalog No. 50-004-50238 Supplier FUJIFILM Biosciences Supplier No. 01712069
L-Ascorbic Acid Phosphate has been shown to promote cell growth and collagen synthesis Specifically Asc 2-P increases the steady state levels of mRNA for type I collagen chains and elevates the transcriptional rates of type I collagen genes Problems exist however with typical ascorbic acid the greatest of which is its short biological half-life Free ascorbic acid is very unstable in solution especially under culture conditions of neutral pH and 37 degrees C L-Ascorbic acid phosphate is much more stable product and can thus be used more easily and effectively Water-soluble high-purity form of Vitamin C phosphate For research use only RUO