Learn More
FUJIFILM Biosciences L-Ascorbic Acid Phosphate Magn
Catalog No. 5000450230
Encompass_Preferred
L-Ascorbic Acid Phosphate has been shown to promote cell growth and collagen synthesis Specifically Asc 2-P increases the steady state levels of mRNA for type I collagen chains and elevates the transcriptional rates of type I collagen genes Problems exist however with typical ascorbic acid the greatest of which is its short biological half-life Free ascorbic acid is very unstable in solution especially under culture conditions of neutral pH and 37 degrees C L-Ascorbic acid phosphate is much more stable product and can thus be used more easily and effectively
Catalog No. 50-004-50230 Supplier FUJIFILM Biosciences Supplier No. 01912063
L-Ascorbic Acid Phosphate has been shown to promote cell growth and collagen synthesis Specifically Asc 2-P increases the steady state levels of mRNA for type I collagen chains and elevates the transcriptional rates of type I collagen genes Problems exist however with typical ascorbic acid the greatest of which is its short biological half-life Free ascorbic acid is very unstable in solution especially under culture conditions of neutral pH and 37 degrees C L-Ascorbic acid phosphate is much more stable product and can thus be used more easily and effectively