Learn More
Sigma Aldrich Fine Chemicals Biosciences L-Ascorbic acid reagent gr500G
Catalog No. 502897337
Encompass
L-Ascorbic acid biosynthetic pathway varies in plants animals and fungi. It is synthesized from L-gulono-1 4-lactone by the action of enzyme L-gulono-1 4-lactone oxidase (GLO).
Catalog No. 50-289-7337 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A7506500G
L-Ascorbic acid biosynthetic pathway varies in plants animals and fungi. It is synthesized from L-gulono-1 4-lactone by the action of enzyme L-gulono-1 4-lactone oxidase (GLO).