Learn More
Sigma Aldrich Fine Chemicals Biosciences L Ascorbic acid reagent gr5KG
Catalog No. 502903133
Encompass
L-Ascorbic acid biosynthetic pathway varies in plants animals and fungi. It is synthesized from L-gulono-1 4-lactone by the action of enzyme L-gulono-1 4-lactone oxidase (GLO).
Catalog No. 50-290-3133 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A75065KG
L-Ascorbic acid biosynthetic pathway varies in plants animals and fungi. It is synthesized from L-gulono-1 4-lactone by the action of enzyme L-gulono-1 4-lactone oxidase (GLO).