Learn More
Medchemexpress LLC L-Ascorbic acid sodium salt | 134-03-2 | 99.0% | 5 G
SDP
Catalog No. 5000406468
Encompass_Preferred
L-Ascorbic acid sodium salt is an electron donor and an endogenous antioxidant agent. It selectively inhibits Cav3.2 channels with an IC50 of 6.5 μM. It also acts as a collagen deposition enhancer and an elastogenesis inhibitor.
  • Functions as an electron donor
  • Acts as an endogenous antioxidant agent
  • Selectively inhibits Cav3.2 channels
  • Enhances collagen deposition
  • Inhibits elastogenesis
Catalog No. 50-004-06468 Supplier Medchemexpress LLC Supplier No. HYB0166A5G
L-Ascorbic acid sodium salt is an electron donor and an endogenous antioxidant agent. It selectively inhibits Cav3.2 channels with an IC50 of 6.5 μM. It also acts as a collagen deposition enhancer and an elastogenesis inhibitor.
  • Functions as an electron donor
  • Acts as an endogenous antioxidant agent
  • Selectively inhibits Cav3.2 channels
  • Enhances collagen deposition
  • Inhibits elastogenesis