Learn More
Medchemexpress LLC L-Ascorbic acid (sodium salt) | 134-03-2 | 99.97% | 10 G
SDP
Catalog No. 5000406467
Encompass_Preferred
L-Ascorbic acid sodium salt is an electron donor and an endogenous antioxidant agent. It selectively inhibits Cav3.2 channels with an IC50 of 6.5 μM. Additionally, it acts as a collagen deposition enhancer and an elastogenesis inhibitor, intended for research use only.
  • Endogenous antioxidant agent
  • Selectively inhibits Cav3.2 channels
  • Enhances collagen deposition
  • Inhibits elastogenesis
  • Available in solid and solution forms for various research needs
  • High purity (99.97%)
Catalog No. 50-004-06467 Supplier Medchemexpress LLC Supplier No. HYB0166A10G
L-Ascorbic acid sodium salt is an electron donor and an endogenous antioxidant agent. It selectively inhibits Cav3.2 channels with an IC50 of 6.5 μM. Additionally, it acts as a collagen deposition enhancer and an elastogenesis inhibitor, intended for research use only.
  • Endogenous antioxidant agent
  • Selectively inhibits Cav3.2 channels
  • Enhances collagen deposition
  • Inhibits elastogenesis
  • Available in solid and solution forms for various research needs
  • High purity (99.97%)