Learn More
Medchemexpress LLC L-Ascorbic acid sodium salt (sodium ascorbate) | 134-03-2 | 99.0% | 1 ML
SDP
Catalog No. 5000407201
Encompass_Preferred
L-Ascorbic acid sodium salt acts as an electron donor and an endogenous antioxidant agent. It also functions as a selective inhibitor of Cav3.2 channels, with an IC50 of 6.5 μM. Additionally, this compound enhances collagen deposition and inhibits elastogenesis.
  • Functions as an electron donor.
  • Acts as an endogenous antioxidant agent.
  • Selectively inhibits Cav3.2 channels.
  • Enhances collagen deposition.
  • Inhibits elastogenesis.
Catalog No. 50-004-07201 Supplier Medchemexpress LLC Supplier No. HYB0166A10MM/1ML
L-Ascorbic acid sodium salt acts as an electron donor and an endogenous antioxidant agent. It also functions as a selective inhibitor of Cav3.2 channels, with an IC50 of 6.5 μM. Additionally, this compound enhances collagen deposition and inhibits elastogenesis.
  • Functions as an electron donor.
  • Acts as an endogenous antioxidant agent.
  • Selectively inhibits Cav3.2 channels.
  • Enhances collagen deposition.
  • Inhibits elastogenesis.