Learn More
Medchemexpress LLC L-Ascorbic acid (Standard) | 50-81-7 | MFCD00064328 | 100.0% | 500 MG
SDP
Catalog No. 5000407203
Encompass_Preferred
L-Ascorbic acid (Standard) is the analytical standard for L-Ascorbic acid, designed for research and analytical applications. It functions as an electron donor and an endogenous antioxidant agent.
  • Analytical standard for research and analytical applications
  • Acts as an electron donor
  • Endogenous antioxidant agent
  • Selectively inhibits Cav3.2 channels with an IC50 of 6.5 μM
  • Enhances collagen deposition
  • Inhibits elastogenesis
  • Exhibits anti-cancer effects through generation of reactive oxygen species (ROS) and selective damage to cancer cells
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-07203 Supplier Medchemexpress LLC Supplier No. HYB0166R500MG
L-Ascorbic acid (Standard) is the analytical standard for L-Ascorbic acid, designed for research and analytical applications. It functions as an electron donor and an endogenous antioxidant agent.
  • Analytical standard for research and analytical applications
  • Acts as an electron donor
  • Endogenous antioxidant agent
  • Selectively inhibits Cav3.2 channels with an IC50 of 6.5 μM
  • Enhances collagen deposition
  • Inhibits elastogenesis
  • Exhibits anti-cancer effects through generation of reactive oxygen species (ROS) and selective damage to cancer cells
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS