Learn More
Medchemexpress LLC L-Ascorbic acid (Standard) | 50-81-7 | MFCD00064328 | 99.97% | 100 MG
SDP
Catalog No. 5000406296
Encompass_Preferred
L-Ascorbic acid (Standard) is an analytical standard of L-Ascorbic acid, intended for research and analytical applications. As an electron donor and endogenous antioxidant agent, it selectively inhibits Cav3.2 channels, enhances collagen deposition, and inhibits elastogenesis. It also demonstrates anti-cancer effects by generating reactive oxygen species (ROS) and selectively damaging cancer cells.
  • Analytical standard
  • Reference standard for qualitative, quantitative, and methodological research
  • Used in HPLC, GC, and MS experiments
  • Electron donor
  • Endogenous antioxidant agent
  • Selectively inhibits Cav3.2 channels with an IC50 of 6.5 μM
  • Collagen deposition enhancer
  • Elastogenesis inhibitor
  • Exhibits anti-cancer effects through ROS generation
Catalog No. 50-004-06296 Supplier Medchemexpress LLC Supplier No. HYB0166R100MG
L-Ascorbic acid (Standard) is an analytical standard of L-Ascorbic acid, intended for research and analytical applications. As an electron donor and endogenous antioxidant agent, it selectively inhibits Cav3.2 channels, enhances collagen deposition, and inhibits elastogenesis. It also demonstrates anti-cancer effects by generating reactive oxygen species (ROS) and selectively damaging cancer cells.
  • Analytical standard
  • Reference standard for qualitative, quantitative, and methodological research
  • Used in HPLC, GC, and MS experiments
  • Electron donor
  • Endogenous antioxidant agent
  • Selectively inhibits Cav3.2 channels with an IC50 of 6.5 μM
  • Collagen deposition enhancer
  • Elastogenesis inhibitor
  • Exhibits anti-cancer effects through ROS generation