Learn More
Sigma Aldrich Fine Chemicals Biosciences L-Ascorbic acid tested acc250G
Catalog No. 502898727
Encompass
L-Ascorbic acid is the most biologically active form of ascorbic acid. It is hydrophilic and an unstable compound. L-Ascorbic acid contains a six-carbon lactone produced by plants and some animal species but not by humans and other primates. Ascorbic acid also referred to as Vitamin C is a water-soluble vitamin. Ascorbic acid carries a neutral charge which gets converted to ascorbate by protonation. Vitamin C is a part of citrus fruits such as broccoli strawberries turnip.
Catalog No. 50-289-8727 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 95212250G
L-Ascorbic acid is the most biologically active form of ascorbic acid. It is hydrophilic and an unstable compound. L-Ascorbic acid contains a six-carbon lactone produced by plants and some animal species but not by humans and other primates. Ascorbic acid also referred to as Vitamin C is a water-soluble vitamin. Ascorbic acid carries a neutral charge which gets converted to ascorbate by protonation. Vitamin C is a part of citrus fruits such as broccoli strawberries turnip.