Learn More
Alkali Scientific L Ascorbic Acid [Vitamin C], 500 G
SDP
Catalog No. 5000660440
Encompass_Preferred
L-Ascorbic Acid [Vitamin C], 500g, is a laboratory-grade reagent used in a variety of biochemical and pharmaceutical applications. Ascorbic acid is a powerful antioxidant that plays a vital role in protecting cells from oxidative damage and is involved in collagen synthesis, immune function, and iron absorption. It is commonly used in cell culture media and as a reducing agent in chemical reactions. In laboratories, it is used for the synthesis of other compounds and as a preservative in food, cosmetic, and pharmaceutical products. This high-quality reagent is suitable for research and manufacturing processes that require accurate formulation and stability in experiments and formulations.
Catalog No. 50-006-60440 Supplier Alkali Scientific Supplier No. ASC001CP
L-Ascorbic Acid [Vitamin C], 500g, is a laboratory-grade reagent used in a variety of biochemical and pharmaceutical applications. Ascorbic acid is a powerful antioxidant that plays a vital role in protecting cells from oxidative damage and is involved in collagen synthesis, immune function, and iron absorption. It is commonly used in cell culture media and as a reducing agent in chemical reactions. In laboratories, it is used for the synthesis of other compounds and as a preservative in food, cosmetic, and pharmaceutical products. This high-quality reagent is suitable for research and manufacturing processes that require accurate formulation and stability in experiments and formulations.