Learn More
Selleck Chemical LLC L-Ascorbyl 6-palmitate 50mg 137-66-6
SDP
Supplier:  Selleck Chemical LLC S253250MG
Encompass_Preferred
Ascorbyl palmitate is an ester formed from ascorbic acid and palmitic acid creating a fat-soluble form of vitamin C. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-594-8