Learn More
Cayman Chemical L-AsparagIn-d3hydrate 10mg
SDP
Catalog No. 502910443
Encompass_Preferred
An internal standard for the quantification of L-asparagine by GC- or LC-MS
Catalog No. 50-291-0443 Supplier Cayman Chemical Supplier No. 3483510 MG
An internal standard for the quantification of L-asparagine by GC- or LC-MS