Learn More
Medchemexpress LLC HY-P1923 5mg , L-Asparaginase CAS:9015-68-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP19235MG
Encompass_Preferred
HY-P1923 5mg L-Asparaginase CAS: 9015-68-3 L-Asparaginase (L-ASNase) is a deamidating enzyme that catalyses the hydrolysis of L-asparagine and L-glutamine, and can be used for the research of acute lymphoblastic leukemia. L-Asparaginase depletes L-asparagine from plasma resulting in inhibition of RNA and DNA synthesis with the subsequent blastic cell apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0329