Learn More
Medchemexpress LLC L-asparagine-13C4,15N2,d3 monohydrate | 2483829-30-5 | 99.9% | 159.11 | 13C4H7D3 15N2O4 | 1 MG
SDP
Catalog No. 5000301781
Encompass_Preferred
L-Asparagine-13C4,15N2,d3 monohydrate is an isotopically labeled L-asparagine (13C, 15N, and deuterium) supplied as the monohydrate. It is intended as a stable-isotope reference standard for mass spectrometry, metabolic tracing, and labeled-peptide synthesis. The material is a white to off-white solid with high isotopic enrichment and high chemical purity.
  • Isotopically labeled for accurate mass spectrometry quantitation.
  • High chemical purity for trace analyses.
  • Supplied as a stable monohydrate solid for reliable handling and storage.
  • Suitable for metabolic tracing and isotopic labeling studies.
  • Available in small analytical quantities for standards and method development.
Catalog No. 50-003-01781 Supplier Medchemexpress LLC Supplier No. HYW017443S21MG
L-Asparagine-13C4,15N2,d3 monohydrate is an isotopically labeled L-asparagine (13C, 15N, and deuterium) supplied as the monohydrate. It is intended as a stable-isotope reference standard for mass spectrometry, metabolic tracing, and labeled-peptide synthesis. The material is a white to off-white solid with high isotopic enrichment and high chemical purity.
  • Isotopically labeled for accurate mass spectrometry quantitation.
  • High chemical purity for trace analyses.
  • Supplied as a stable monohydrate solid for reliable handling and storage.
  • Suitable for metabolic tracing and isotopic labeling studies.
  • Available in small analytical quantities for standards and method development.