Learn More
Medchemexpress LLC L-asparagine-13C4,15N2,d3 monohydrate | 2483829-30-5 | 99.9% | 159.11 g/mol | 13C4H7D3 15N2O4 | 10 MG
SDP
Catalog No. 5000299978
Encompass_Preferred
Isotopically labeled L-asparagine monohydrate supplied as a small research pack for stable isotope tracing, metabolic studies, and use as an analytical internal standard. The sample contains 13C, 15N, and deuterium labels and is provided with purity and COA documentation.
  • Isotopically labeled for precise metabolic tracing.
  • Suitable as a mass spectrometry internal standard.
  • High purity for analytical applications.
  • Supplied as a small, accurately weighed research pack.
  • Accompanied by COA, SDS, and handling documentation.
Catalog No. 50-002-99978 Supplier Medchemexpress LLC Supplier No. HYW017443S210MG
Isotopically labeled L-asparagine monohydrate supplied as a small research pack for stable isotope tracing, metabolic studies, and use as an analytical internal standard. The sample contains 13C, 15N, and deuterium labels and is provided with purity and COA documentation.
  • Isotopically labeled for precise metabolic tracing.
  • Suitable as a mass spectrometry internal standard.
  • High purity for analytical applications.
  • Supplied as a small, accurately weighed research pack.
  • Accompanied by COA, SDS, and handling documentation.