Learn More
Medchemexpress LLC L-asparagine-13C4,15N2 monohydrate | 202406-87-9 | MFCD01073445 | 98.0% | 153.07 | 13C4H7 15N2O4 | 5 MG
SDP
Catalog No. 5000301108
Encompass_Preferred
L-asparagine-13C4,15N2 monohydrate is an isotopically labeled amino acid provided as a monohydrate for use as a stable-isotope internal standard and tracer in mass spectrometry, metabolite tracing, and isotope labeling studies. It contains four 13C labels and two 15N labels and is manufactured to analytical-grade purity for reliable quantitative work.
  • Isotopically labeled with 13C4 and 15N2 for unambiguous mass shifts.
  • High purity suitable for analytical and quantitative applications.
  • Provided as a monohydrate solid for straightforward handling and storage.
  • Intended for use as an internal standard in mass spectrometry workflows.
  • Available in small-scale quantities for method development and validation.
Catalog No. 50-003-01108 Supplier Medchemexpress LLC Supplier No. HYW017443S5MG
L-asparagine-13C4,15N2 monohydrate is an isotopically labeled amino acid provided as a monohydrate for use as a stable-isotope internal standard and tracer in mass spectrometry, metabolite tracing, and isotope labeling studies. It contains four 13C labels and two 15N labels and is manufactured to analytical-grade purity for reliable quantitative work.
  • Isotopically labeled with 13C4 and 15N2 for unambiguous mass shifts.
  • High purity suitable for analytical and quantitative applications.
  • Provided as a monohydrate solid for straightforward handling and storage.
  • Intended for use as an internal standard in mass spectrometry workflows.
  • Available in small-scale quantities for method development and validation.