Learn More
Medchemexpress LLC L-asparagine-13C4,15N2 monohydrate | 202406-87-9 | MFCD01073445 | 98.0% | 153.07 g/mol | 13C4H7 15N2O4 | 10 MG
SDP
Catalog No. 5000301411
Encompass_Preferred
L-Asparagine-13C4,15N2 monohydrate is an isotopically labeled form of the amino acid L-asparagine (monohydrate) intended for use as a tracer or internal standard in quantitative analysis, metabolic flux experiments, and mass spectrometry. CAS 202406-87-9; molecular weight 153.07 g/mol.
  • Isotopically labeled at four carbon positions (13C) and two nitrogen positions (15N).
  • Supplied as a solid monohydrate for ease of handling.
  • High chemical purity (98.0%) suitable for analytical applications.
  • Optimized for use as a tracer and internal standard in mass spectrometry.
  • Available in small, mg-scale pack sizes for research and method development.
  • Compatible with common sample preparation workflows for quantitative assays.
Catalog No. 50-003-01411 Supplier Medchemexpress LLC Supplier No. HYW017443S10MG
L-Asparagine-13C4,15N2 monohydrate is an isotopically labeled form of the amino acid L-asparagine (monohydrate) intended for use as a tracer or internal standard in quantitative analysis, metabolic flux experiments, and mass spectrometry. CAS 202406-87-9; molecular weight 153.07 g/mol.
  • Isotopically labeled at four carbon positions (13C) and two nitrogen positions (15N).
  • Supplied as a solid monohydrate for ease of handling.
  • High chemical purity (98.0%) suitable for analytical applications.
  • Optimized for use as a tracer and internal standard in mass spectrometry.
  • Available in small, mg-scale pack sizes for research and method development.
  • Compatible with common sample preparation workflows for quantitative assays.