Learn More
Medchemexpress LLC L-Asparagine-13C4 m 1mg
SDP
Catalog No. 5000669517
Encompass_Preferred
L-Asparagine-13C4 monohydrate is the 13C-labeled L-Asparagine monohydrate (HY-W017443) L-Asparagine monohydrate is an essential amino acid for leukemic cells and a substrate for L-Asparaginase L-Asparaginase is a potent anti-leukemic enzyme that promotes asparagine (Asn) and glutamine (Gln) depletion and inhibits protein biosynthesis in lymphoblasts Removal of L-asparagine from plasma by L-Asparaginase results in inhibition of RNA and DNA synthesis and subsequent apoptosis L-Asparaginase has cell-killing ability in vitro and in vivo and selectively inhibits the growth of cancer cells with low asparagine synthetase (AASNS) expression L-Asparagine monohydrate can be used as a biomarker and sensor for the study of childhood acute lymphoblastic leukemia[1][2][3][4][5][6]
Catalog No. 50-006-69517 Supplier Medchemexpress LLC Supplier No. 1MGHYN0667S3
L-Asparagine-13C4 monohydrate is the 13C-labeled L-Asparagine monohydrate (HY-W017443) L-Asparagine monohydrate is an essential amino acid for leukemic cells and a substrate for L-Asparaginase L-Asparaginase is a potent anti-leukemic enzyme that promotes asparagine (Asn) and glutamine (Gln) depletion and inhibits protein biosynthesis in lymphoblasts Removal of L-asparagine from plasma by L-Asparaginase results in inhibition of RNA and DNA synthesis and subsequent apoptosis L-Asparaginase has cell-killing ability in vitro and in vivo and selectively inhibits the growth of cancer cells with low asparagine synthetase (AASNS) expression L-Asparagine monohydrate can be used as a biomarker and sensor for the study of childhood acute lymphoblastic leukemia[1][2][3][4][5][6]