Learn More
Medchemexpress LLC L-asparagine-15N2 monohydrate | 287484-32-6 | MFCD01073515 | 98.0% | 152.12 | C4H10(15N)2O4 | 10 MG
SDP
Catalog No. 5000295925
Encompass_Preferred
L-Asparagine-15N2 monohydrate is the 15N2-isotope labeled form of the amino acid L-asparagine supplied as a monohydrate. It is intended for use as an internal standard or labeled reagent in mass spectrometry, metabolomics, and tracer studies; reported molecular weight is 152.12 and CAS number is 287484-32-6.
  • Isotope-labeled (15N2) amino acid for internal standard use.
  • Monohydrate form, supplied as a solid suitable for dissolution or storage under inert atmosphere.
  • High purity (98.0% by NMR) for analytical applications.
  • Suitable for mass spectrometry, metabolomics, and tracer studies.
  • Store solid at 4°C, protect from light, and store under nitrogen.
Catalog No. 50-002-95925 Supplier Medchemexpress LLC Supplier No. HYN0667S210MG
L-Asparagine-15N2 monohydrate is the 15N2-isotope labeled form of the amino acid L-asparagine supplied as a monohydrate. It is intended for use as an internal standard or labeled reagent in mass spectrometry, metabolomics, and tracer studies; reported molecular weight is 152.12 and CAS number is 287484-32-6.
  • Isotope-labeled (15N2) amino acid for internal standard use.
  • Monohydrate form, supplied as a solid suitable for dissolution or storage under inert atmosphere.
  • High purity (98.0% by NMR) for analytical applications.
  • Suitable for mass spectrometry, metabolomics, and tracer studies.
  • Store solid at 4°C, protect from light, and store under nitrogen.