Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Asparagine, 99% [Anhydrous], MP Biomedicals

Catalog No. ICN19463090 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19463090 500 g
ICN19463025 25 g
ICN19463080 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19463090 Supplier MP Biomedicals, Inc Supplier No. 0219463090
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 70-47-3
Molecular Formula C4H8N2O3
Molecular Weight (g/mol) 132.119
InChI Key DCXYFEDJOCDNAF-REOHCLBHSA-N
Synonym l-asparagine, asparagine, s-asparagine, aspartamic acid, agedoite, altheine, h-asn-oh, crystal vi, asparamide, asparagine acid
PubChem CID 6267
ChEBI CHEBI:17196
IUPAC Name (2S)-2,4-diamino-4-oxobutanoic acid
SMILES C(C(C(=O)O)N)C(=O)N

Specifications

Quantity 500 g
Formula Weight 132.1
Percent Purity 99%
Grade Cell Culture
Physical Form Crystals
Chemical Name or Material L-Asparagine, 99% [Anhydrous]
Recommended Storage Room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.