Learn More
Medchemexpress LLC L-asparagine-amide-15N monohydrate | 204451-47-8 | MFCD01075622 | 99.8% | 151.13 g/mol | C4H10N2O4 | 10 MG
SDP
Catalog No. 5000301740
Encompass_Preferred
L-asparagine-amide-15N monohydrate is a 15N stable isotope-labeled derivative of L-asparagine amide supplied as a monohydrate. It is intended for use as an internal standard and tracer in mass spectrometry, metabolic labeling, and analytical studies that require isotopic enrichment. The material is provided as a high-purity, white to off-white solid suitable for research and analytical applications.
  • 15N isotopic labeling for tracer and analytical studies.
  • High purity suitable for reference standards and quantitative analysis.
  • Monohydrate solid form for stable handling and storage.
  • Compatible with mass spectrometry and metabolic profiling workflows.
  • Small pack sizes for analytical and method development use.
Catalog No. 50-003-01740 Supplier Medchemexpress LLC Supplier No. HYW017443S110MG
L-asparagine-amide-15N monohydrate is a 15N stable isotope-labeled derivative of L-asparagine amide supplied as a monohydrate. It is intended for use as an internal standard and tracer in mass spectrometry, metabolic labeling, and analytical studies that require isotopic enrichment. The material is provided as a high-purity, white to off-white solid suitable for research and analytical applications.
  • 15N isotopic labeling for tracer and analytical studies.
  • High purity suitable for reference standards and quantitative analysis.
  • Monohydrate solid form for stable handling and storage.
  • Compatible with mass spectrometry and metabolic profiling workflows.
  • Small pack sizes for analytical and method development use.