Learn More
Creative Biomart Asparaginase Escherichia coli
Supplier:  Creative Biomart THP0056
Encompass_Preferred
Asparaginase derived from Escherichia coli (L-asparagine amidohydrolase, EC 3.5.1.1) is an enzyme responsible for the metabolism of L-asparagine, by catalyzing L-asparagine into L-aspartic acid and ammonia. It also facilitates the production of oxaloacetate which is needed for general cellular metabolism.
Catalog No. 50-208-4107