Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Asparagine Anhydrous MP Biomedicals

Catalog No. ICN10079480 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10079480 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10079480 Supplier MP Biomedicals, Inc Supplier No. 0210079480
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Anhydrous chemical for biochemical and laboratory use

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70-47-3
Molecular Formula C4H8N2O3
Molecular Weight (g/mol) 132.119
InChI Key DCXYFEDJOCDNAF-REOHCLBHSA-N
Synonym l-asparagine, asparagine, s-asparagine, aspartamic acid, agedoite, altheine, h-asn-oh, crystal vi, asparamide, asparagine acid
PubChem CID 6267
ChEBI CHEBI:17196
IUPAC Name (2S)-2,4-diamino-4-oxobutanoic acid
SMILES C(C(C(=O)O)N)C(=O)N

Specifications

Quantity 100 g
Formula Weight 132.1
Grade Biochemical
Chemical Name or Material L-Asparagine, Anhydrous
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.