Learn More
Research Products International Corp L-Asparagine, Monohydrate, 100 Grams
SDP
Supplier:  Research Products International Corp A50030100.0
Encompass_Preferred
L-Asparagine Monohydrate is commonly used as a component in cell culture media. It provides a source of nitrogen for cell growth and is particularly important for the cultivation of certain cell types, especially in the production of recombinant proteins and antibodies. 
As an amino acid, it is involved in protein synthesis, contributing to the structure and function of various proteins within cells.
L-Asparagine Monohydrate is utilized in research, particularly in studies related to cell biology, molecular biology, biochemistry and is valuable tool for understanding cellular processes.
Catalog No. 50-213-149