Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-asparagine monohydrate | 5794-13-8 | MFCD00151038 | 99.8% | 150.13 g/mol | C4H10N2O4 | 1 G
SDP

Catalog No. 5000301381
Encompass_Preferred

L-asparagine monohydrate is the monohydrate form of the amino acid L-asparagine, supplied as a white to off-white solid. It is commonly used as a biochemical reagent and analytical standard, and serves as a substrate in assays involving L-asparaginase. The material is intended for research applications that require a high-purity amino acid standard.

  • High purity: 99.8%.
  • White to off-white solid appearance.
  • Chemical formula: C4H10N2O4; molecular weight: 150.13 g/mol.
  • Used as a biochemical reagent and analytical standard.
  • Available in small packaging suitable for research: 1 g, 5 g, 10 g.

Catalog No. 50-003-01381 Supplier Medchemexpress LLC Supplier No. HYW0174431G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-asparagine monohydrate is the monohydrate form of the amino acid L-asparagine, supplied as a white to off-white solid. It is commonly used as a biochemical reagent and analytical standard, and serves as a substrate in assays involving L-asparaginase. The material is intended for research applications that require a high-purity amino acid standard.

  • High purity: 99.8%.
  • White to off-white solid appearance.
  • Chemical formula: C4H10N2O4; molecular weight: 150.13 g/mol.
  • Used as a biochemical reagent and analytical standard.
  • Available in small packaging suitable for research: 1 g, 5 g, 10 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.