Learn More
Bioworld L-Asparagine, Monohydrate, 100 g
Supplier:  Bioworld 401002841
Encompass_Preferred
Synonym: (S)-2-Aminosuccinic acid 4-amide, (S)-(+)-2-Aminosuccinamic acid, L-Aspartic acid 4-amide Aproteinogenic amino acidUsed in biomanufacturing cell culture systems for the production of therapeutic recombinant proteins and monoclonal antibodies and in cell culture based research.Appearance: SolidDensity : 1.543 g/cm3HS Code : 29241000Melting Point: 234-238°C
Catalog No. 50-255-288