Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals L(+)-Asparagine monohydrate, 99%

Catalog No. AC175270010
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC175270010 1 kg Glass Bottle
AC175271000 100 g Glass Bottle
AC175272500 250 g Glass Bottle
AC175270050 5 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC175270010 Supplier Thermo Scientific Chemicals Supplier No. 175270010
Add to Cart

L(+)-Asparagine monohydrate, 99%, C4H10N2O4, CAS Number-5794-13-8, asparagine monohydrate, l +-asparagine monohydrate, asparagine, asparagine nf, l-asparagine hydrate, h-asn-oh.h2o, unii-2pd79vf521, l-asparagine monohydrate, l-asparagine, monohydrate, s-2,4-diamino-4-oxobutanoic acid hydrate, 250g | CAS: 5794-13-8 | C4H10N2O4 | 150.134 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5794-13-8
Molecular Formula C4H10N2O4
Molecular Weight (g/mol) 150.134
MDL Number MFCD00151038
InChI Key RBMGJIZCEWRQES-DKWTVANSSA-N
Synonym l-asparagine monohydrate, l +-asparagine monohydrate, l-asparagine hydrate, asparagine monohydrate, h-asn-oh.h2o, l-asparagine, monohydrate, asparagine nf, s-2,4-diamino-4-oxobutanoic acid hydrate, asparagine, unii-2pd79vf521
PubChem CID 170358
IUPAC Name (2S)-2,4-diamino-4-oxobutanoic acid;hydrate
SMILES C(C(C(=O)O)N)C(=O)N.O

Specifications

Melting Point 233.0°C to 235.0°C
Ammonium 1000ppm max.
Arsenic (As) 1ppm max.
Color White
Chlorine (Cl) 210ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Assay Percent Range 99%
Loss on Drying 11.5% to 12.5% (130°C, 3 hrs)
Other Amino Acids 1% max. (TLC)
Packaging Glass Bottle
Linear Formula H2NCOCH2CH(NH2)CO2H·H2O
Specific Rotation + 35.00
Sulfate 280ppm max.
Transmittance 98% min. (c=2, H2O, 430nm) 1cm cell
Sulfated Ash 0.1% max.
Solubility Information Solubility in water: 30g/L water (20°C). Other solubilities: soluble in acids and alkalies, practically insoluble in methanol, ethanol, ether, and benzene
Specific Rotation Condition + 35.00 (20.00°C c=10,6N HCl)
Quantity 1 kg
Formula Weight 150.14
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material L(+)-Asparagine monohydrate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.