Learn More
Medchemexpress LLC L-α-Asparagine, N-acetyl-L-tyrosyl-L-valyl-L-alanyl-N-(4-methyl-2-oxo-2H-1-benzopyran- | 149231-65-2 | 99.6% | 665.69 | 25 MG
SDP
Catalog No. 5000397314
Encompass_Preferred
Ac-YVAD-AMC is a potent caspase inhibitor, commonly used in research to study programmed cell death (apoptosis) pathways. It acts as a fluorogenic substrate for caspase-1, caspase-4, and caspase-5, enabling the detection and quantification of their enzymatic activity in various biological assays.
  • Potent caspase inhibitor
  • Useful in apoptosis research
  • Fluorogenic substrate for caspase-1, caspase-4, and caspase-5
  • Enables detection and quantification of caspase activity
Catalog No. 50-003-97314 Supplier Medchemexpress LLC Supplier No. 25MGHYP2717
Ac-YVAD-AMC is a potent caspase inhibitor, commonly used in research to study programmed cell death (apoptosis) pathways. It acts as a fluorogenic substrate for caspase-1, caspase-4, and caspase-5, enabling the detection and quantification of their enzymatic activity in various biological assays.
  • Potent caspase inhibitor
  • Useful in apoptosis research
  • Fluorogenic substrate for caspase-1, caspase-4, and caspase-5
  • Enables detection and quantification of caspase activity