Learn More
Cayman Chemical L-Asparagine (hydrate), Purity: ≥95%, CAS: 5794-13-8, 1 g
SDP
Catalog No. 502949962
Encompass_Preferred
A non-essential amino acid; reverses ASNS siRNA knockdown-induced decreases in proliferation in a panel of six cancer cell lines at 0.3 mM
Catalog No. 50-294-9962 Supplier Cayman Chemical Supplier No. 345191 G
A non-essential amino acid; reverses ASNS siRNA knockdown-induced decreases in proliferation in a panel of six cancer cell lines at 0.3 mM