Learn More
Cayman Chemical L-Asparagine (hydrate), Purity: ≥95%, CAS: 5794-13-8, 5 g
SDP
Catalog No. 502950564
Encompass_Preferred
A non-essential amino acid; reverses ASNS siRNA knockdown-induced decreases in proliferation in a panel of six cancer cell lines at 0.3 mM
Catalog No. 50-295-0564 Supplier Cayman Chemical Supplier No. 345195 G
A non-essential amino acid; reverses ASNS siRNA knockdown-induced decreases in proliferation in a panel of six cancer cell lines at 0.3 mM