Learn More
Medchemexpress LLC L-aspartamide, 1-acetyl-L-prolyl-L-histidyl-L-seryl-L-cysteinyl-, mono(trifluoroacetate) (salt) | 904763-27-5 | 99.0% | C25H36F3N9O10S | 10MG
SDP
Catalog No. 5000259932
Encompass_Preferred
ATN-161 trifluoroacetate salt is a peptide integrin α5β1 antagonist supplied as the trifluoroacetate (TFA) salt for research use. It has been reported to inhibit angiogenesis and reduce growth of liver metastases in murine models. The product is supplied as a white to off-white solid with high purity and accompanying analytical documentation.
  • Peptide integrin α5β1 antagonist, suitable for preclinical research.
  • Provided as the trifluoroacetate salt for improved stability and handling.
  • High purity (approximately 99.0%), with COA and analytical data available.
  • Solid form, white to off-white, convenient for storage and formulation.
  • Storage recommendations: refrigerated sealed storage; long-term in solvent at -80°C.
  • Available in small-milligram quantities for laboratory experiments.
Catalog No. 50-002-59932 Supplier Medchemexpress LLC Supplier No. HY13535A10MG
ATN-161 trifluoroacetate salt is a peptide integrin α5β1 antagonist supplied as the trifluoroacetate (TFA) salt for research use. It has been reported to inhibit angiogenesis and reduce growth of liver metastases in murine models. The product is supplied as a white to off-white solid with high purity and accompanying analytical documentation.
  • Peptide integrin α5β1 antagonist, suitable for preclinical research.
  • Provided as the trifluoroacetate salt for improved stability and handling.
  • High purity (approximately 99.0%), with COA and analytical data available.
  • Solid form, white to off-white, convenient for storage and formulation.
  • Storage recommendations: refrigerated sealed storage; long-term in solvent at -80°C.
  • Available in small-milligram quantities for laboratory experiments.