Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-aspartic acid-13C4,15N | 202468-27-7 | MFCD00144566 | 98.0% | 138.07 | 13C4H7 15NO4 | 10 MG
SDP

Catalog No. 5000296380
Encompass_Preferred

L-Aspartic acid-13C4,15N is a stable isotope-labeled form of the amino acid L-aspartic acid used as a reference standard and tracer in analytical, metabolic, and mass spectrometry applications.

  • Stable isotope-labeled (13C4, 15N) suitable as an internal standard for LC-MS/MS and quantitative assays.
  • High purity; ≥98.0%.
  • Molecular weight 138.07 provides consistent mass spectrometric behavior.
  • Chemical formula 13C4H7 15NO4 reflects isotopic labeling.
  • Supplied in small laboratory quantities for analytical workflows.

Catalog No. 50-002-96380 Supplier Medchemexpress LLC Supplier No. HYN0666S110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Aspartic acid-13C4,15N is a stable isotope-labeled form of the amino acid L-aspartic acid used as a reference standard and tracer in analytical, metabolic, and mass spectrometry applications.

  • Stable isotope-labeled (13C4, 15N) suitable as an internal standard for LC-MS/MS and quantitative assays.
  • High purity; ≥98.0%.
  • Molecular weight 138.07 provides consistent mass spectrometric behavior.
  • Chemical formula 13C4H7 15NO4 reflects isotopic labeling.
  • Supplied in small laboratory quantities for analytical workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.