Learn More
Medchemexpress LLC L-Aspartic acid-15N | 3715-16-0 | 134.10 | 50 MG
SDP
Catalog No. 5000388752
Encompass_Preferred
L-Aspartic acid-15N is the 15N-labeled version of L-Aspartic acid, an amino acid that can penetrate the blood-brain barrier. It is commonly used for preparing prodrugs to target colon and cecal tissues and in the study of inflammatory conditions.
  • Used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Intended for research use only
Catalog No. 50-003-88752 Supplier Medchemexpress LLC Supplier No. HYN0666S350MG
L-Aspartic acid-15N is the 15N-labeled version of L-Aspartic acid, an amino acid that can penetrate the blood-brain barrier. It is commonly used for preparing prodrugs to target colon and cecal tissues and in the study of inflammatory conditions.
  • Used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Intended for research use only