Learn More
Medchemexpress LLC L-Aspartic acid | 56-84-8 | MFCD00002616 | 133.10 | 25 G
SDP
Catalog No. 5000389458
Encompass_Preferred
L-Aspartic acid is an amino acid commonly used for preparing prodrugs to target colon and cecal tissues. It is also frequently utilized in the study of inflammatory conditions and is classified as an endogenous metabolite.
  • Amino acid
  • Commonly used for preparing prodrugs to target colon and cecal tissues
  • Used in the study of inflammatory conditions
  • Endogenous metabolite
Catalog No. 50-003-89458 Supplier Medchemexpress LLC Supplier No. 25GHYN0666
L-Aspartic acid is an amino acid commonly used for preparing prodrugs to target colon and cecal tissues. It is also frequently utilized in the study of inflammatory conditions and is classified as an endogenous metabolite.
  • Amino acid
  • Commonly used for preparing prodrugs to target colon and cecal tissues
  • Used in the study of inflammatory conditions
  • Endogenous metabolite