Learn More
Medchemexpress LLC L-Aspartic acid-d3 | 3842-25-9 | 136.12 | 25 MG
SDP
Catalog No. 5000389445
Encompass_Preferred
L-Aspartic acid-d3 is a deuterium labeled L-Aspartic acid, an amino acid that can penetrate the blood-brain barrier. It is used in preparing prodrugs for colon and cecal tissues and in studying inflammatory conditions. This compound also serves as a tracer and internal standard for quantitative analysis by methods like NMR, GC-MS, or LC-MS.
  • Deuterium labeled amino acid.
  • Penetrates the blood-brain barrier.
  • Used for prodrug preparation and inflammatory condition studies.
  • Functions as a tracer and internal standard.
  • Suitable for NMR, GC-MS, or LC-MS quantitative analysis.
  • Purity of 99.9%.
  • Appears as a white to off-white solid.
Catalog No. 50-003-89445 Supplier Medchemexpress LLC Supplier No. 25MGHYN0666S8
Add to Cart
Add to Cart
L-Aspartic acid-d3 is a deuterium labeled L-Aspartic acid, an amino acid that can penetrate the blood-brain barrier. It is used in preparing prodrugs for colon and cecal tissues and in studying inflammatory conditions. This compound also serves as a tracer and internal standard for quantitative analysis by methods like NMR, GC-MS, or LC-MS.
  • Deuterium labeled amino acid.
  • Penetrates the blood-brain barrier.
  • Used for prodrug preparation and inflammatory condition studies.
  • Functions as a tracer and internal standard.
  • Suitable for NMR, GC-MS, or LC-MS quantitative analysis.
  • Purity of 99.9%.
  • Appears as a white to off-white solid.