Learn More
AJINOMOTO USA INC L-Aspartic Acid (L-Asp) 1Kg
Supplier:  AJINOMOTO USA INC 32572
Encompass
Chemical Name: (S)-Aminosuccinic Acid; Chemical Formula: C4H7NO4; Molecular Weight: 133.1000; Nitrogen Content: 0.10520000; CAS Number: 56-84-8; Description: White crystals or crystalline powder 1Kg
Catalog No. NC1383825