Learn More
Medchemexpress LLC L-Aspartic acid, N-[9-(5-O-phosphono-β-D-ribofuranosyl)-9H-purin-6-yl]- | 19046-78-7 | 95.0% | 1 MG
SDP
Catalog No. 5000341369
Encompass_Preferred
Adenylosuccinic acid, also known as Adenylosuccinate or Aspartyl adenylate, is a purine ribonucleoside monophosphate that plays a vital role as a nucleotide cycle metabolite. This compound can be converted into fumaric acid through the action of adenylosuccinate lyase. Research indicates that Adenylosuccinic acid holds potential for the study of Duchenne Muscular Dystrophy (DMD).
  • Adenylosuccinic acid is a purine ribonucleoside monophosphate.
  • It functions as a nucleotide cycle metabolite.
  • It can be converted to fumaric acid by adenylosuccinate lyase.
  • It shows potential for research into Duchenne Muscular Dystrophy (DMD).
Catalog No. 50-003-41369 Supplier Medchemexpress LLC Supplier No. HY1271371MG
Add to Cart
Add to Cart
Adenylosuccinic acid, also known as Adenylosuccinate or Aspartyl adenylate, is a purine ribonucleoside monophosphate that plays a vital role as a nucleotide cycle metabolite. This compound can be converted into fumaric acid through the action of adenylosuccinate lyase. Research indicates that Adenylosuccinic acid holds potential for the study of Duchenne Muscular Dystrophy (DMD).
  • Adenylosuccinic acid is a purine ribonucleoside monophosphate.
  • It functions as a nucleotide cycle metabolite.
  • It can be converted to fumaric acid by adenylosuccinate lyase.
  • It shows potential for research into Duchenne Muscular Dystrophy (DMD).