Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Aspartic Acid Sodium Salt, Monohydrate, 98-101%, Spectrum™ Chemical
SDP

Catalog No. 18603732 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
12 kg
50 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-732 50 kg Poly Bottle
18-603-729 25 g Poly Bottle
18-603-730 100 g Poly Bottle
18-603-731 1 kg Poly Bottle
18-603-733 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-603-732 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A137550KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

A1375, 323194-76-9, C4H6NO4Na·H2O

Spectrum™ Chemical L-Aspartic Acid Sodium Salt, Monohydrate

Chemical Identifiers

CAS 323194-76-9
Molecular Formula C4H8NNaO5
Molecular Weight (g/mol) 173.10
MDL Number MFCD00152960
InChI Key PPTHNBYUFXSJPS-UHFFFAOYNA-M
IUPAC Name sodium 2-amino-3-carboxypropanoate hydrate
SMILES O.[Na+].NC(CC(O)=O)C([O-])=O

Specifications

CAS Max % 100%
Physical Form Crystalline Powder or Needles
Quantity 50 kg
Percent Purity 98 to 101%
Grade Ungraded
Loss on Drying 0.3%
Packaging Poly Bottle
Chemical Name or Material L-Aspartic Acid Sodium Salt, Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.