Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Aspartic Acid, Spectrum™ Chemical
SDP

Catalog No. 18620205 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-205 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-620-205 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. AS12525KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Versatile organic intermediate used for the preparation of culture media.

  • Physical State: Solid (Crystals, Crystalline powder)
  • Chemical Formula: C4H7NO4
  • Molecular Weight: 133.10
  • Assay: 98.5 %
  • Melting Point: 270°C to 271°C
  • Specific Rotation ([α]D20): 24.5°
  • Solubility: 1 g in 222.2 mL water at 20°C; Insoluble in ethanol
  • Stability: Stable at normal conditions; Light sensitive
  • Recommended Use: Preparation of culture media; Organic intermediate
  • Quality Standards: ISO 9001:2015; Operates under cGMP

Chemical Identifiers

CAS 56-84-8
Molecular Weight (g/mol) 133.10

Specifications

Packaging Drum
Quantity 25 kg
Percent Purity 98.50%
Chemical Name or Material L-Aspartic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.